ELISA Blocker Blocking Buffer, Thermo Scientific, Thermo Scientific

Supplier: 0000013509

N502
PIN502EA 385.2 USD
PIN502
ELISA Blocker Blocking Buffer, Thermo Scientific, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers

ELISA Blocker Blocking Buffer is a protein buffer that effectively blocks plate binding sites that remain in polystyrene microplate wells following initial coating steps in ELISA procedures. The ready-to-use buffer is optimized and routinely used for sandwich ELISA procedures involving cytokines, such as many of the Thermo Scientific Pierce ELISA Kits.


  • Store at 2–8˚C
  • For Research Use Only
  • Not for use in diagnostic procedures


Features OF THE ELISA Blocking Buffer:


Size: 1L (sufficient for thirty 96-well plates)
Application: Use as a blocking buffer in ELISA. The buffer is ready to use. No dilution is required.
Instructions for use in an ELISA: Following the incubation with a coating antibody, aspirate the coating antibody from the plate and add 300 µL of ELISA Blocker Blocking Buffer to each well. Cover the plate with a plate sealer and incubate for a minimum of 1 hour at room temperature (22-25˚C). Aspirate blocking buffer and proceed with the ELISA procedure or dry the plate for future use.
Plate Drying: Plates should be dried uncovered, at room temperature, for a minimum of 1 hour. For optimal drying, plates should not be stacked and humidity should be at 30-40%. If plates are stacked, or humidity is high, the drying time should be increased. Once drying is complete, seal each plate in a foil pouch with a desiccant. When sealed with a desiccant, plates may be stored at 2-8˚C for up to 6 months.


Size: 1L (sufficient for thirty 96-well plates)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR