Pierce™ High Capacity Endotoxin Removal Resin, Thermo Scientific

Supplier: 0000013509

88270 88271 88272
PI88270EA 505.38 USD
PI88270 PI88271 PI88272
Pierce™ High Capacity Endotoxin Removal Resin, Thermo Scientific
Protein Purification Protein Purification Resins

Pierce™ High Capacity Endotoxin Removal Resins are ideal for the removal of endotoxins from protein or antibody samples prior to cell transfection or animal injection.


  • Bind up to 2 000 000 EU/ml to eliminate <99% of endotoxins
  • Recover greater than 85% of protein sample


Pierce™ High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000 EU/ml binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/ml. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/ml. This resin is offered in a slurry format for custom packing of endotoxin removal columns for gravity flow or flow rates ranging from 10 to 15 ml/hour.


Delivery information: Supplied as 50% slurry in 20% ethanol


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR