Ethidium bromide solution 0,625 mg/mL, Pierce™

Supplier: LIFE TECHNOLOGIES MS

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

17896
PI17896EA 112.92 USD
PI17896
Ethidium bromide solution 0,625 mg/mL, Pierce™
Ethidium bromide

Thermo Scientific Pierce Ethidium Bromide is available as a dilute and concentrated solution to accommodate most molecular biology applications for fluorescent DNA visualization.


Ethidium Bromide is an intercalating dye used in the visualization of DNA and RNA in electrophoresis gels and cesium chloride density gradients.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Boiling Pt: 72
MDL Number: MFCD00011724
CAS Number: 1239-45-8
Merck Index: 13,04751

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR