Pierce™ North2South™ Hybridization Stringency Wash Buffer (2X), Thermo Scientific
Supplier: 0000013509
37555
PI-37555EA 287.69 USD
PI-37555
Pierce™ North2South™ Hybridization Stringency Wash Buffer (2X), Thermo Scientific
Buffers Electrophoresis Buffers Hybridization Buffers
Thermo Scientific North2South Hybridization Stringency Wash Buffer (2X), for use with the North2South Chemiluminescent Hybridization and Detection Kit.
  • pH 6.9
  • Aqueous buffer
Caution: For Research Use Only. Not for use in diagnostic procedures. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8˚C.