CR100® Sterile Latex Gloves, Cardinal
Supplier: Cardinal Health
2Y1624 2Y1640 2Y1625 2Y1626 2Y1627 2Y1628 2Y1629 2Y1630
PH2Y1624CS 756.59 USD
PH2Y1624 PH2Y1640 PH2Y1625 PH2Y1626 PH2Y1627 PH2Y1628 PH2Y1629 PH2Y1630
CR100® Sterile Latex Gloves, Cardinal
Gloves
Versatile, high-quality latex gloves can be used in critical environments that demand stringent aseptic procedures.
  • Sterile, clean processed, powder free
  • Comfortable flex-and-stretch design
  • Anti-slip, textured finish enhances the grip for better handling
  • Beaded cuff with reinforced ribs prevents constriction and reduces rolldown
Hand-specific and powder-free. Low particulates and extractables. Bisque finish, tapered cuff. Sterilized by gamma radiation to a Sterility Assurance Level of 10⁻⁶ in accordance with AAMI guidelines. Color: natural.