One-Way Luer-Lok Stopcocks for Nucleic Acid Purification Systems, Promega®
Supplier: Promega Corporation
A7261
PAA7261EA 17.38 USD
PAA7261
One-Way Luer-Lok Stopcocks for Nucleic Acid Purification Systems, Promega®
Stopcocks
One-Way Luer-Lok stopcocks vacuum manifold for nucleic acid purification system. Spinlock adapters on the columns from the Wizard DNA purifications systems can be attached to the upper row of stopcocks for easy manipulation of the stopcock valves.