Secondary Pressure Vessels System, Saf-T-Pak
Supplier: SAF-T-PAK INC MS
STP-714 STP-710 STP-730 STP700 STP-740 STP-701
MSPP-STP714CS 546.15 USD
MSPP-STP714 MSPP-STP-710 MSPP-STP-730 MSPP-STP700 MSPP-STP-740 MSPP-STP701
Secondary Pressure Vessels System, Saf-T-Pak
Bags Sample Bags
Secondary pressure vessels are pressure tested to hold an internal pressure differential of 95 kPa without leaking. The product is a two-part system consisting of an inner outer envelope they are stronger than today’s present technology.
  • Biohazard and leakproof poly bag
  • Outer envelope container of Tyvek
  • Each unit is for single use and is disposable
Infectious shipments: This system is the tested part of the shipperand restricted to that shipper.
Diagnostic shipments: this system is readily available for any specimen shipment using any of theMore Product Information