Echotherm™ Full Featured Digital and Stirring Hot Plates with Solid Ceramic or Milled Flat Cast Aluminum Heater Tops
Supplier: TORREY PINES SCIENTIFIC MS
HP30A-2
MSPP-HP30A-2EA 1525.54 USD
MSPP-HP30A-2
Echotherm™ Full Featured Digital and Stirring Hot Plates with Solid Ceramic or Milled Flat Cast Aluminum Heater Tops
Heaters Hot Plates
These hot plates are designed to be the easiest to use and most accurate fully featured hot plates in the world. Temperature measurement of a solution is accurate to 1% over the entire temperature range using a 100 ohm platinum RTD sensor and measurement for precision and accuracy.
  • Extremely easy use by anyone in your lab
  • Solid ceramic heater top for quick heating on a flat, white, chemically resistant surface are framed in cast aluminum to keep the edges cool
  • Milled flat cast aluminum tops available for heating directly on the heater surface
  • PID
More Product Information