Small Animal Ear Tags, Braintree Scientific
Supplier: BRAINTREE SCIENTIFIC INC MS
EP-1001-2000
MSPP-EP1001200EA 265.85 USD
MSPP-EP1001200
Small Animal Ear Tags, Braintree Scientific
Animal Supplies Animal ID Tags and Punches
These stainless steel applicators are designed to seal tags with minimal pressure, while providing a secure seal.
  • Applicator can be used with or without the spring
Spring applies slight closing pressure to help hold tag in applicator prior to sealing.