Vi-CELL™ MetaFLEX Sensor Cassette, Beckman Coulter
Supplier: Beckman Coulter
B93030
MSPP-B93030EA 3410.17 USD
MSPP-B93030
Vi-CELL™ MetaFLEX Sensor Cassette, Beckman Coulter
Cell Counters and Flow Cytometers Electronic Cell Counters
The Vi-CELL MetaFLEX sensor cassette SC300 measures pH, pO2, pCO2, Glucose, Lactate, Electrolytes and more parameters associated with whole blood analysis.
For use with the Vi-CELL MetaFLEX™ bioanalyte analyser.
Order Now
SPECIFICATIONS
Environmentally Preferable