Air Science USA® Purair® Filters, Air Science USA LLC
Supplier: Air Science
ASTS-001 ASTS-012 ASTS-013 ASTS-015 ASTS-030 ASTS-PRF (6 PACK)
MSPP-ASTS-001EA 294.74 USD
MSPP-ASTS-001 MSPP-ASTS-012 MSPP-ASTS-013 MSPP-ASTS-015 MSPP-ASTS-030 MSPP-ASTSPRF
Air Science USA® Purair® Filters, Air Science USA LLC
Fume Hoods and Enclosures Ductless Fume Hoods
Purair® carbon filters for use with ductless fume hoods.
  • Fit Air Science Purair® Basic ductless fume hoods
  • Easy to install
Purair® filters for Basic ductless fume hoods for models P5-24, P5-36 (one is required), for the larger P5-48 models (two filters are required).