Multichannel Reagent Reservoirs and Bases, Integra
Supplier: 0000018277
4312 4332 4322 4304 4305 4306
MSPP-4312CS 229.23 USD
MSPP-4312 MSPP-4332 MSPP-4322 MSPP-4304 MSPP-4305 MSPP-4306
Multichannel Reagent Reservoirs and Bases, Integra
Reagent Reservoirs
These polystyrene reagent reservoirs offer a convenient solution for the temporary storage of non-solvent based liquids during any pipetting application.
  • Integrated volume graduations result in more accurate measurements and less reagent waste
  • Convenient pour back spout is molded into each reagent reservoir, allowing precise control of liquid flow
  • Multichannel reagent reservoirs feature an extra deep trough that accumulates liquid, reducing dead volumes and ensuring maximum reagent recovery
Each reusable reservoir base securely holds disposable clearMore Product Information