Onyx PCX Derivatization Instruments
Supplier: Pickering Labs
1155-1051 1155-1096 1155-1061 1155-1071 1155-1081 1155-1011 1155-1021 1155-1099 1155-1103 1155-1105 1155-1031 1155-1101
MSPP-1155-1051EA 48615.38 USD
MSPP-1155-1051 MSPP-1155-1096 MSPP-1155-1061 MSPP-1155-1071 MSPP-1155-1081 MSPP-1155-1011 MSPP-1155-1021 MSPP-1155-1099 MSPP-1155-1103 MSPP-1155-1105 MSPP-1155-1031 MSPP-1155-1101
Onyx PCX Derivatization Instruments
Chromatography Systems Derivatization Systems
Pickering Laboratories offers the only instrumentation optimized for the analysis of amino acids, carbamates, glyphosate, mycotoxins, antibiotics and many other applications. Each component is specifically designed to enhance sensitivity and selectivity. Only Pickering Laboratories offers complete application support, including chemicals, columns, methods and post-column systems.
  • True pulse-free syringe pump with single piece ceramic barrels
  • Programmable flow rates of 50 to 1500 μl/minute
  • Refill cycle of 60 seconds or less
  • Automatic piston wash, reagent
More Product Information