Water, AR® ACS, deionized, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Dihydrogen oxide

M.O.C. Category NI

H453-05 H453-07 H453-09
MKH45305CS 231.63 USD
MKH45305 MKH45307 MKH45309
Water, AR® ACS, deionized, Macron Fine Chemicals™
Water

Distilled water quality, deionized water.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Chloride (Cl) ≤0.4 ppm
ACS - Electrical Resistivity at 25°C (Mohm.cm) ≥0.5
ACS - Heavy Metals (as Pb) ≤0.01 ppm
ACS - Nitrate (NO₃) ≤0.4 ppm
ACS - Phosphate (PO₄) ≤1.0 ppm
ACS - Residue after Evaporation ≤0.0002 %
ACS - Silicate (as SiO₂) ≤0.01 ppm
ACS - Specific Conductance at 25°C. (/uohms⁻¹ cm⁻¹) ≤2.0
ACS - Substances Reducing Permanganate Passes Test
ACS - Sulfate (SO₄) ≤1.0 ppm
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR