Oxalic acid 10% w/v aqueous solution, StandARd® (APHA), Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ethanedioic acid

M.O.C. Category N

H281-05
MKH28105CS 637.95 USD
MKH28105
Oxalic acid 10% w/v aqueous solution, StandARd® (APHA), Macron Fine Chemicals™
Oxalic acid
Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365
Density: 1.02 g/cm³
MDL Number: MFCD00002573
CAS Number: 144-62-7
UN: 3265
ADR: 8 III
Merck Index: 13,06980

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (H₂C₂O₄·2H₂O)(g/100 mL) 9.5 - 10.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR