Perchloric acid 70%, AR Select® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

8828-14
MK882814CS 4514.71 USD
MK882814
Perchloric acid 70%, AR Select® ACS, Macron Fine Chemicals™
Perchloric acid
Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 39
Melting Pt: -101
Density: 1.53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HClO₄) 69.0 - 72.0 %
ACS - Chloride (Cl) ≤ 0.001 %
ACS - Color (APHA) ≤ 10
ACS - Heavy Metals (as Pb) ≤ 0.0001 %
ACS - Nitrogen Compounds (as N) ≤ 0.001 %
ACS - Residue after Ignition ≤ 0.003 %
ACS - Silicate and Phosphate (as SiO₂) ≤ 0.0005 %
ACS - Sulfate (SO₄) ≤ 0.001 %
Determine by Flame Photometry & ICP - Aluminum (Al) ≤ 100 ppb
Determine by Flame Photometry & ICP - Barium (Ba) ≤ 100 ppb
Determine by Flame Photometry & ICP - Beryllium (Be) ≤ 10 ppb
Determine by Flame Photometry & ICP - Bismuth (Bi) ≤ 10 ppb
Determine by Flame Photometry & ICP - Boron (B) ≤ 1000 ppb
Determine by Flame Photometry & ICP - Cadmium (Cd) ≤ 50 ppb
Determine by Flame Photometry & ICP - Calcium (Ca) ≤ 1000 ppb
Determine by Flame Photometry & ICP - Chromium (Cr) ≤ 100 ppb
Determine by Flame Photometry & ICP - Cobalt (Co) ≤ 50 ppb
Determine by Flame Photometry & ICP - Copper (Cu) ≤ 100 ppb
Determine by Flame Photometry & ICP - Gallium (Ga) ≤ 500 ppb
Determine by Flame Photometry & ICP - Germanium (Ge) ≤ 500 ppb
Determine by Flame Photometry & ICP - Gold (Au) ≤ 500 ppb
Determine by Flame Photometry & ICP - Iron (Fe) ≤ 100 ppb
Determine by Flame Photometry & ICP - Lead (Pb) ≤ 100 ppb
Determine by Flame Photometry & ICP - Lithium (Li) ≤ 500 ppb
Determine by Flame Photometry & ICP - Magnesium (Mg) ≤ 100 ppb
Determine by Flame Photometry & ICP - Manganese (Mn) ≤ 50 ppb
Determine by Flame Photometry & ICP - Mercury (Hg) ≤ 100 ppb
Determine by Flame Photometry & ICP - Molybdenum (Mo) ≤ 10 ppb
Determine by Flame Photometry & ICP - Nickel (Ni) ≤ 100 ppb
Determine by Flame Photometry & ICP - Potassium (K) ≤ 500 ppb
Determine by Flame Photometry & ICP - Silicon (Si) ≤ 500 ppb
Determine by Flame Photometry & ICP - Silver (Ag) ≤ 100 ppb
Determine by Flame Photometry & ICP - Sodium (Na) ≤ 1000 ppb
Determine by Flame Photometry & ICP - Strontium (Sr) ≤ 100 ppb
Determine by Flame Photometry & ICP - Thallium (Tl) ≤ 50 ppb
Determine by Flame Photometry & ICP - Tin (Sn) ≤ 500 ppb
Determine by Flame Photometry & ICP - Vanadium (V) ≤ 10 ppb
Determine by Flame Photometry & ICP - Zinc (Zn) ≤ 100 ppb
Determine by Flame Photometry & ICP - Zirconium (Zr) ≤ 10 ppb
Determine by Flame Photometry & ICP
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR