Dichloromethane 99,5 GC_ASSAY_METHOD, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Methane dichloride, Methylene chloride, Methylene chloride

M.O.C. Category N

4881-08 4881-19 4881-06 4881-04 4881-26
MK488108CS 1877.71 USD
MK488108 MK488119 MK488106 MK488104 MK488126
Dichloromethane 99,5 GC_ASSAY_METHOD, AR® ACS, Macron Fine Chemicals™
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: -95
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₂Cl₂) (by GC) ≥99.5 %
Appearance (clear, colorless liquid) Passes Test
ACS - Carbon Tetrachloride (mg/L) ≤10.0000 ppm
ACS - Color (APHA) ≤10
ACS - Free Halogens Passes Test
ACS - Residue after Evaporation ≤0.002 %
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Water (H₂O)(by coulometry) ≤0.02 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR