Calcium hydroxide 95.0-100.5% (by EDTA titration), powder FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Slaked lime

Management of Change (MOC) category = RL

4188-25 4188-40 4188-24 MACR4188-24 MACR4188-05
MK418825EA 1011.7 USD
MK418825 MK418840 JT4188-05 JT4188-24 MK4188-24 MK4188-05
Calcium hydroxide 95.0-100.5% (by EDTA titration), powder FCC, Macron Fine Chemicals™
Calcium hydroxide
Formula: Ca(OH)₂
MW: 74.09 g/mol
Melting Pt: 550
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8 III
Merck Index: 13,01675

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Assay (Ca(OH)₂) (by EDTA titrn) 95.0 - 100.5 %
Appearance Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Carbonate (CO₃) Passes Test
FCC - Acid-Insoluble Substances ≤0.5 %
FCC - Arsenic (As) ≤3 ppm
FCC - Lead (Pb) ≤2 ppm
FCC - Fluoride (F) ≤0.005 %
FCC - Magnesium and Alkali Salts ≤4.8 %
Thru U.S. No. 325 Sieve, For Information Only
Barium (Ba), For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR