Trichloroacetic acid (TCA) 99,0 acidimetry on anhydrous substance, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

M.O.C. Category N

2928-10 2928-08
MK292810CS 1290.49 USD
MK292810 MK292808
Trichloroacetic acid (TCA) 99,0 acidimetry on anhydrous substance, crystals, AR® ACS, Macron Fine Chemicals™
Trichloroacetic acid (TCA)

Crystals.

Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 196
Melting Pt: 54…56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 1839
ADR: 8 II
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Cl₃CCOOH) (dried basis)(by acidimetry) ≥99.0 %
ACS - Clarity of Solution Passes Test
ACS - Insoluble Matter ≤0.01 %
ACS - Residue after Ignition ≤0.03 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Nitrate (NO₃) ≤0.002 %
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Sulfate (SO₄) ≤0.02 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Iron (Fe) ≤0.001 %
ACS - Substances Darkened by H₂SO₄ Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR