meta-Phosphoric acid 33,5 - 36,5% Stabilised, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Metaphosphoric acid

M.O.C. Category N

2816-04 MACR2816-06 2816-06
MK281604CS 11637.13 USD
MK281604 BAMK281606COV MK281606
meta-Phosphoric acid 33,5 - 36,5% Stabilised, AR® ACS, Macron Fine Chemicals™
meta-Phosphoric acid
Formula: (HPO₃)n
MW: 79.98 g/mol
Boiling Pt: 260
Melting Pt: 21
Density: 2.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011351
CAS Number: 37267-86-0
EINECS: 253-433-4
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HPO₃) 33.5 - 36.5 %
ACS - Appearance (colorless, small glassy scales) Passes Test
ACS - Stabilizer (as NaPO₃) 57.0 - 63.0 %
ACS - Arsenic (As) ≤0.0001 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.005 %
ACS - Iron (Fe) ≤0.005 %
ACS - Nitrate (NO₃) ≤0.001 %
Solubility Passes Test
ACS - Substances Reducing Permanganate (as H₃PO₃) ≤0.02 %
ACS - Sulfate (SO₄) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR