Masterflex® Transfer Tubing, PharMed® BPT, Avantor®
Supplier: VWR International
MFLX96880-05 MFLX96880-10 MFLX96880-14 MFLX96880-00 MFLX96880-02 MFLX96880-03 MFLX96880-04 MFLX96880-06 MFLX96880-08 MFLX96880-12 MFLX96880-15 MFLX96880-16 MFLX96880-18 MFLX96880-20 MFLX96880-22 MFLX96880-30
MFLX96880-05EA 218 USD
MFLX96880-05 MFLX96880-10 MFLX96880-14 MFLX96880-00 MFLX96880-02 MFLX96880-03 MFLX96880-04 MFLX96880-06 MFLX96880-08 MFLX96880-12 MFLX96880-15 MFLX96880-16 MFLX96880-18 MFLX96880-20 MFLX96880-22 MFLX96880-30
Masterflex® Transfer Tubing, PharMed® BPT, Avantor®
Tubing Transfer Tubing
Protect your light-sensitive samples.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. Made according to GMP. Temperature range is -51 to 270 °More Product Information