Masterflex® Transfer Tubing, Flexible Vinyl, Avantor®
Supplier: VWR International
MFLX96480-00 MFLX96480-01 MFLX96480-02 MFLX96480-04 MFLX96480-05 MFLX96480-06 MFLX96480-07 MFLX96480-08 MFLX96480-09 MFLX96480-10 MFLX96480-11 MFLX96480-13 MFLX96480-14 MFLX96480-15 MFLX96480-18 MFLX96480-19 MFLX96480-03 MFLX96480-12 MFLX96480-16 MFLX96480-17 MFLX96480-21
MFLX96480-00EA 23 USD
MFLX96480-00 MFLX96480-01 MFLX96480-02 MFLX96480-04 MFLX96480-05 MFLX96480-06 MFLX96480-07 MFLX96480-08 MFLX96480-09 MFLX96480-10 MFLX96480-11 MFLX96480-13 MFLX96480-14 MFLX96480-15 MFLX96480-18 MFLX96480-19 MFLX96480-03 MFLX96480-12 MFLX96480-16 MFLX96480-17 MFLX96480-21
Masterflex® Transfer Tubing, Flexible Vinyl, Avantor®
Tubing Transfer Tubing
Ideal for general-purpose applications.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
A good general-purpose tubing material, vinyl tubing (also known as PVC tubing) offers good chemical resistance. Its transparency allows easy visual identification of flow through the tube. This vinyl tubing's good abrasionMore Product Information