Masterflex® Single-Use I/P® Gamma-Irradiated Precision Pump Tubing, Avantor®
Supplier: VWR International
MFLX96119-73 MFLX96119-82 MFLX96123-73 MFLX96122-73 MFLX96122-82 MFLX96123-82
MFLX96119-73EA 466.25 USD
MFLX96119-73 MFLX96119-82 MFLX96123-73 MFLX96122-73 MFLX96122-82 MFLX96123-82
Masterflex® Single-Use I/P® Gamma-Irradiated Precision Pump Tubing, Avantor®
Tubing Peristaltic Pump Tubing
Reduce the risk of microbial contamination.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Gamma sensitive labeling confirms treatment
Masterflex® gamma-More Product Information