Masterflex® Transfer Tubing, Versilon™ C-544-A IB, Avantor®
Supplier: VWR International
MFLX95637-02 MFLX95637-00 MFLX95637-01 MFLX95637-04 MFLX95637-05 MFLX95637-06 MFLX95637-07
MFLX95637-02EA 807 USD
MFLX95637-02 MFLX95637-00 MFLX95637-01 MFLX95637-04 MFLX95637-05 MFLX95637-06 MFLX95637-07
Masterflex® Transfer Tubing, Versilon™ C-544-A IB, Avantor®
Tubing Transfer Tubing
No plasticizers.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The Versilon™ C-544-A IB ether-based polyurethane pressure tubing, is ideal for abrasive and viscous slurry transfer, instrumentation control lines, food and cosmetic processing, and coolant recovery systems. Characteristics include exceptionalMore Product Information