Masterflex® Transfer Tubing, Tygon® F-4040-A, Fuel and Lubricant, Avantor®
Supplier: VWR International
MFLX95633-00 MFLX95633-01 MFLX95633-02 MFLX95633-03 MFLX95633-04 MFLX95633-05 MFLX95633-08 MFLX95633-09 MFLX95633-10 MFLX95633-06
MFLX95633-00EA 88.5 USD
MFLX95633-00 MFLX95633-01 MFLX95633-02 MFLX95633-03 MFLX95633-04 MFLX95633-05 MFLX95633-08 MFLX95633-09 MFLX95633-10 MFLX95633-06
Masterflex® Transfer Tubing, Tygon® F-4040-A, Fuel and Lubricant, Avantor®
Tubing Transfer Tubing
No swelling or embrittlement.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The Tygon tubing is ideal for fuels and industrial lubricants—gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants. Resists most hydrocarbons, embrittlement and swelling, ozone- and UV-resistant, with low-More Product Information