Masterflex® I/P® Rapid-Load® Pump Heads for Precision and High-Performance Precision Tubing
Supplier: Avantor Fluid Handling
77603-22 77603-20
MFLX77603-22EA 3410 USD
MFLX77603-22 MFLX77603-20
Masterflex® I/P® Rapid-Load® Pump Heads for Precision and High-Performance Precision Tubing
Pumps Pump Heads Peristaltic Pump Heads
Built for bioprocess applications - designed for ease of use and process flexibility.
  • Single-action opens pump head and occlusion surface - so tubing changes are fast and easy
  • Hard-coat anodized aluminum construction is extremely durable and corrosion resistant - ready to withstand washdown environments and common sanitizing solutions
  • Stainless steel (SS) rotor/roller assembly for durability and chemical resistance
  • Guide rollers stabilize tubing and ensure quiet operation
  • Efficient operational footprint saves bench space and provides installation
More Product Information