4 to 20 mA Pace Pumps for High-Viscosity Applications
Supplier: EQUIP SOLUTIONS
LVF4MA-VTT5
MFLX74147-10EA 1740 USD
MFLX74147-10 MFLX74147-00 MFLX74147-20 MFLX74147-30 MFLX74147-40
4 to 20 mA Pace Pumps for High-Viscosity Applications
Pumps Diaphragm Pumps
Reliable metering performance – guided dual check valves provide precise seating and excellent priming and suction lift.
  • Constructed of PVC, PTFE, and 316 Stainless Steel
High-viscosity 4 to 20 mA pumps handle viscous fluids up to 20,000 cp. Pumps feature automatic control with 4 to 20 mA DC direct or external pacing, reliable timing circuits, stop function, and manually adjustable stroke rate and length. Operate pump in automatic or manual mode by selecting the proper setting on the "Auto-Off-Manual" switch. Circuit protection guards against voltageMore Product Information