Masterflex® Digital Benchtop Gear Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
74014-02 74014-12 74014-22 74014-32 74014-42 74014-52 74014-62 74014-72 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 07002-25 75211-70 & 07002-14 75211-70 & 74012-11 75211-70 & 74012-21 75211-70 & 74012-31 75211-70 & 07003-04 75211-70 & 74012-51 75211-70 & 74012-61 75211-70 & 07002-25
MFLX74014-02EA 3910 USD
MFLX74014-02 MFLX74014-12 MFLX74014-22 MFLX74014-32 MFLX74014-42 MFLX74014-52 MFLX74014-62 MFLX74014-72 MFLX74014-02-AU MFLX74014-12-AU MFLX74014-22-AU MFLX74014-32-AU MFLX74014-42-AU MFLX74014-52-AU MFLX74014-62-AU MFLX74014-72-AU MFLX74014-02-DK MFLX74014-12-DK MFLX74014-22-DK MFLX74014-32-DK MFLX74014-42-DK MFLX74014-52-DK MFLX74014-62-DK MFLX74014-72-DK MFLX74014-02-IN MFLX74014-12-IN MFLX74014-22-IN MFLX74014-32-IN MFLX74014-42-IN MFLX74014-52-IN MFLX74014-62-IN MFLX74014-72-IN MFLX74014-02-IL MFLX74014-12-IL MFLX74014-22-IL MFLX74014-32-IL MFLX74014-42-IL MFLX74014-52-IL MFLX74014-62-IL MFLX74014-72-IL MFLX74014-02-EU MFLX74014-12-EU MFLX74014-22-EU MFLX74014-32-EU MFLX74014-42-EU MFLX74014-52-EU MFLX74014-62-EU MFLX74014-72-EU MFLX74014-02-UK MFLX74014-12-UK MFLX74014-22-UK MFLX74014-32-UK MFLX74014-42-UK MFLX74014-52-UK MFLX74014-62-UK MFLX74014-72-UK MFLX74014-02-CH MFLX74014-12-CH MFLX74014-22-CH MFLX74014-32-CH MFLX74014-42-CH MFLX74014-52-CH MFLX74014-62-CH MFLX74014-72-CH MFLX74014-02-IT MFLX74014-12-IT MFLX74014-22-IT MFLX74014-32-IT MFLX74014-42-IT MFLX74014-52-IT MFLX74014-62-IT MFLX74014-72-IT
Masterflex® Digital Benchtop Gear Pump Systems, Avantor®
Pumps Gear Pumps
Order the complete pump drive and head system.
  • Graphical LCD shows four operating modes
  • IP33 rating ensures splash and dust resistance
  • Systems deliver pulse-free flow in combination with Micropump® A-mount head
  • Remote control via DB25 female connector on drive
Calibrate, copy, and dispense features produce precise, pulseless batching and dispensing. View pump performance continually. Retrieve displacement and flow values by pump head size.
These precision systems are easily operated from the intuitive, multilanguage LCD front panel or optional footMore Product Information