Micropump® Suction Shoe Gear Pumps for NEMA 56 C Motors
Supplier: MICROPUMP INC
81376+81356 81377 (GC-M35.PVS.E) L19053 L19381+83400 L18413 MFLX07003-41 MFLX73005-05 MFLX73005-07 MFLX73005-09
MFLX07003-32EA 2550 USD
MFLX07003-32 MFLX07003-33 MFLX73005-08 MFLX73005-13 MFLX73005-02 MFLX07003-41 MFLX73005-05 MFLX73005-07 MFLX73005-09
Micropump® Suction Shoe Gear Pumps for NEMA 56 C Motors
Pumps Gear Pumps
Easily attach your gear pump to a motor.
  • Suction shoe design gives higher delivery pressures and superior priming performance.
  • Smooth, pulseless flow
  • Precision, leak-free delivery of fluids
  • Compact and easy to service
  • Use with any NEMA 56 C motors
Magnetically driven, precision-geared pump with unique suction shoe design provides smooth, accurate, pulseless fluid delivery. The magnetic drive, Viton® fluoroelastomer O-rings, and PTFE seal keep fluids securely inside and potential contaminants out. Chemically resistant to a wide range of materials.More Product Information