PuraLev® LeviFlow Single-Use Pump Series
Supplier: LeviFlow
100-91114 100-91115 100-91310 100-91118 100-91311 100-91120
MFLX72072-00EA 3310 USD
MFLX72072-00 MFLX72072-04 MFLX72072-10 MFLX72072-12 MFLX72072-14 MFLX72072-16
PuraLev® LeviFlow Single-Use Pump Series
Pumps Centrifugal Pumps
Safely handle sensitive biotech applications.
  • Single-use disposable pump head reduces risk of contamination
  • Bayonet socket design makes exchanging pump heads quick and easy
  • Absence of narrow gaps between impeller and pump casing eliminates bacteria buildup
The PuraLev single-use magnetic drive centrifugal pump kits feature a low shear-force design ideal for pumping shear-sensitive liquids and cells in biotech and pharmaceutical applications. The stand-alone systems include a drive with a PLC interface, and single-use pump head. The PLC interfaceMore Product Information