Female Cam to Male NPT Threaded Couplers, Polypropylene
Supplier: VWR International
MFLX34020-32 MFLX34020-35 MFLX34020-36
MFLX34020-32EA 16.6 USD
MFLX34020-32 MFLX34020-35 MFLX34020-36
Female Cam to Male NPT Threaded Couplers, Polypropylene
Fittings and Connectors
Cam couplers provide quick disconnection from system.
  • All fittings have EPDM gaskets
  • Constructed of durable polypropylene
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.