Masterflex® Mass Flowmeter Controllers for Gas, Avantor®
Supplier: ALICAT SCIENTIFIC INC
MC-1SCCM-D/5M MC-5SCCM-D/5M MC-10SCCM-D/5M MC-50SCCM-D/5M MC-100SCCM-D/5M MC-200SCCM-D/5M MC-500SCCM-D/5M MC-1SLPM-D/5M MC-5SLPM-D/5M MC-10SLPM-D/5M MC-50SLPM-D/5M MC-100SLPM-D/5M MC-250SLPM-D/5M MCR-500SLPM-D/5M MFLX32907-81
MFLX32907-51EA 2770 USD
MFLX32907-51 MFLX32907-53 MFLX32907-55 MFLX32907-57 MFLX32907-59 MFLX32907-61 MFLX32907-63 MFLX32907-67 MFLX32907-69 MFLX32907-71 MFLX32907-73 MFLX32907-75 MFLX32907-77 MFLX32907-79 MFLX32907-81
Masterflex® Mass Flowmeter Controllers for Gas, Avantor®
Flow Switches
Measure 130 standard gases—user selectable from display.
  • Accuracy of ±0.8% of reading ±0.2% full-scale; repeatability of ±0.2%
  • Measure 30 standard gases—user selectable from display
  • 100 to 1 turndown with ranges of 1 SCCM full-scale up to 1000 SLPM
  • Models with 4 to 20 mA output or totalizer are available
These meters measure flow via pressure drop across a laminar flow element (LFE). Because the flow element makes the flow stream laminar, placement in the process does not require straight pipe runs upstream or downstream of the meter, greatlyMore Product Information