Kobold® Paddle Type Metal Flow Switches
Supplier: KOBOLD INSTRUMENTS INC
PSR-5115 PSR-5120 PSR-5125 PSR-5132 PSR-5205 PSR-5215 PSR-5225 PSR-5140 PSR-5210 MFLX32481-29 MFLX32481-30 MFLX32481-37 MFLX32481-40
MFLX32481-31EA 207 USD
MFLX32481-31 MFLX32481-32 MFLX32481-33 MFLX32481-34 MFLX32481-35 MFLX32481-36 MFLX32481-38 MFLX32481-41 MFLX32481-42 MFLX32481-29 MFLX32481-30 MFLX32481-37 MFLX32481-40
Kobold® Paddle Type Metal Flow Switches
Flow Switches
Adjust switch point to a maximum of 1450/3600 psi.
  • Switch point adjustable
  • Easy to install
  • Brass or Stainless Steel construction
  • Maximum pressure: 1450/3600 PSI
The device operates as follows: The flowing medium presses against the paddle for the flow switch. The paddle is fitted to one end of a balance arm which is in direct contact with a prestressed leaf spring. At the other end of the balance arm is a permanent magnet. This magnet actuates a reed contact located within a moveable housing outside the media. The reed contact switches on or offMore Product Information