Masterflex® Union Fittings, Hose Barb with Sanitary Clamp, Tee, Avantor®
Supplier: VWR International
MFLX31203-33
MFLX31203-33EA 90.4 USD
MFLX31203-33
Masterflex® Union Fittings, Hose Barb with Sanitary Clamp, Tee, Avantor®
Fittings and Connectors
Sanitary fitting with built-in diaphragm makes sanitary installation easier than ever.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for
More Product Information