Masterflex® Tuf-Steel® Sanitary Screen Tri-Clamp Gaskets, Avantor®
Supplier: VWR International
40MPGTSS200-10SS 40MPGTSS150-20SS 40MPGTSS200-20SS 40MPGTSS150-40SS 40MPGTSS150-60SS 42MPGTSS050-20SS 42MPGTSS050-40SS 40MPGTSS100-40SS 40MPGTSS200-40SS 40MPGTSS200-60SS 40MPGTSS100-100&10SS 40MPGTSS150-100&10SS 40MPGTSS200-100&10SS
MFLX30539-08EA 200 USD
MFLX30539-08 MFLX30539-26 MFLX30539-28 MFLX30539-46 MFLX30539-66 MFLX30539-20 MFLX30539-40 MFLX30539-44 MFLX30539-48 MFLX30539-68 MFLX30539-84 MFLX30539-86 MFLX30539-88
Masterflex® Tuf-Steel® Sanitary Screen Tri-Clamp Gaskets, Avantor®
Seals Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derivedMore Product Information