Masterflex® Strainer Mesh for Low-Cost In-Line Strainer Systems, Avantor®
Supplier: SPECIALTY MFG CO
5580310 (10486) 5580320 (10485) 5580330 (10496) 5730270 (13274) 5580640 (14986) 5730360 (10505) 5730340 (10507) 5730330 (10511) 5580400 (10522) 5580410 (10523) 5580420 (10524) 5730370 (10526) 5730380 (10527)
MFLX29595-31EA 17 USD
MFLX29595-31 MFLX29595-33 MFLX29595-35 MFLX29595-39 MFLX29595-41 MFLX29595-43 MFLX29595-45 MFLX29595-47 MFLX29595-49 MFLX29595-51 MFLX29595-53 MFLX29595-55 MFLX29595-57
Masterflex® Strainer Mesh for Low-Cost In-Line Strainer Systems, Avantor®
Filters Strainers
Easily customize strainer system to suit your application.
  • Choose multiple mesh and micron sizes options based on required filtration area
  • Constructed of 304 stainless steel
Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate, and large. Modify or change components of the same class to meet the demands of your application.
Low-Profile Strainers are ideal for small-scale low-volume filtration. Use in research applications, cleaning equipment, and laboratoryMore Product Information