Masterflex® Transfer Tubing, Polytetrafluoroethylene (PTFE), Avantor®
Supplier: VWR International
MFLX06407-41 MFLX06605-27 MFLX06605-28 MFLX06605-29 MFLX06605-30 MFLX06605-31 MFLX06605-32 MFLX06605-33 MFLX06605-34 MFLX06605-35 MFLX06605-36 MFLX06605-37 MFLX06605-38 MFLX06605-39 MFLX06605-40 MFLX06605-41 MFLX06605-42 MFLX06605-44 MFLX06605-45 MFLX06605-46 MFLX06605-48 MFLX06605-49 MFLX06605-50 MFLX06605-51
MFLX06407-41EA 77.3 USD
MFLX06407-41 MFLX06605-27 MFLX06605-28 MFLX06605-29 MFLX06605-30 MFLX06605-31 MFLX06605-32 MFLX06605-33 MFLX06605-34 MFLX06605-35 MFLX06605-36 MFLX06605-37 MFLX06605-38 MFLX06605-39 MFLX06605-40 MFLX06605-41 MFLX06605-42 MFLX06605-44 MFLX06605-45 MFLX06605-46 MFLX06605-48 MFLX06605-49 MFLX06605-50 MFLX06605-51
Masterflex® Transfer Tubing, Polytetrafluoroethylene (PTFE), Avantor®
Tubing Transfer Tubing
Won't crack under heat or pressure.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures.
Characteristics: Chemically inert, nontoxic, andMore Product Information