Masterflex® Transfer Tubing, Vinyl, Avantor®
Supplier: VWR International
MFLX06405-02 MFLX06405-27 MFLX06405-01 MFLX06405-03 MFLX06405-04 MFLX06405-06 MFLX06405-07 MFLX06405-08 MFLX06405-09 MFLX06405-10 MFLX06405-12 MFLX06405-15 MFLX06405-16 MFLX06405-18 MFLX06405-19 MFLX06405-20 MFLX06405-31 MFLX06405-36 MFLX06405-25 MFLX06405-46
MFLX06405-02EA 34.5 USD
MFLX06405-02 MFLX06405-27 MFLX06405-01 MFLX06405-03 MFLX06405-04 MFLX06405-06 MFLX06405-07 MFLX06405-08 MFLX06405-09 MFLX06405-10 MFLX06405-12 MFLX06405-15 MFLX06405-16 MFLX06405-18 MFLX06405-19 MFLX06405-20 MFLX06405-31 MFLX06405-36 MFLX06405-25 MFLX06405-46
Masterflex® Transfer Tubing, Vinyl, Avantor®
Tubing Transfer Tubing
Ideal for general-purpose applications.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The tubing is ideal for general-purpose lab use. Characteristics include fair chemical and abrasion resistance, odorless, nontoxic, and transparent. Tubing is manufactured according to GMP and has a temperature range of -45More Product Information