KONTES® Quartz Beaker, Kimble®
Supplier: DWK Life Sciences (KIMBLE)
319011-0050
KT319011-0050EA 230.49 USD
KT319011-0050
KONTES® Quartz Beaker, Kimble®
Beakers
This quartz beaker/crucible is designed for muffle furnace ashing procedures.
  • Useful where chemical purity is a factor in subsequent analysis
  • Quartz will also satisfy high temperature requirements to 1050°C
  • Very useful for determining trace metals in environmental samples
The cover has to be ordered separately.