Polysorbate 80, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate

M.O.C. Category N

X257-09 X257-07 X257-01
JTX257-9CS 11096.45 USD
JTX257-9 JTX257-7 JTX257-01
Polysorbate 80, J.T.Baker®
Polysorbate 80

Polyoxyethylene (20) Sorbitan Monooleate

Melting Pt: 100
Flash Pt: 113
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-018-3

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test
Not intended for Injectable Drug Use.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR