Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99,8 Dry_basis, BAKER ANANYZED™ ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

M.O.C. Category N

X171-07 X171-05 X171-03
JTX171-7EA 407.96 USD
JTX171-7 JTX171-5 JTX171-3
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99,8 Dry_basis, BAKER ANANYZED™ ACS
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Lot analysis on label.

Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 357
Melting Pt: 169…172 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₄H₁₁NO₃) (dried basis) 99.8 - 100.1 %
Absorbance of a 40% Solution: Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤2 %
Insoluble Matter ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
pH of 0.1M Solution at 25°C, For Information Only
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR