Sodium hexametaphosphate, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium metaphosphate, SHMP

Management of Change (MOC) category = N

V030-09
JTV030-9EA 247.89 USD
JTV030-9
Sodium hexametaphosphate, J.T.Baker®
Sodium hexametaphosphate
Formula: Na₆P₆O₁₈
Melting Pt: 610 °C
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00136045
CAS Number: 10124-56-8
EINECS: 233-343-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as P₂O₅) ≥65.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR