Ammonia solution 28,0 - 30,0% NH3 basis, BAKER ANALYZED® A.C.S. Reagent ACS zA

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

M.O.C. Category NI

9721-03 9721-05 9721-02 9721-06 9721-00 9721-01 9721-15 9721-33 BAKR9721-00
JT9721-3CS 466.04 USD
JT9721-3 JT9721-5 JT9721-2 JT9721-6 JT9721-0 JT9721-1 JT9721-15 JT9721-33 SCJT9721-0 SCJT9721-1 SCJT9721-33 JT9721-18
Ammonia solution 28,0 - 30,0% NH3 basis, BAKER ANALYZED® A.C.S. Reagent ACS zA
Ammonia solution

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.88…0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For laboratory, research or manufacturing use.
Appearance (colorless and free from suspended matter or sediment) Passes Test
Assay (as NH₃) 28.0 - 30.0%
Color (APHA) ≤5
Specific Gravity at 60/60 °F 0.896 - 0.902
Residue after Ignition ≤0.0020%
Carbon Dioxide (CO₂) ≤0.002%
Substances Reducing Permanganate Passes Test
Chloride (Cl) ≤0.5 ppm
Nitrate (NO₃) ≤2 ppm
Phosphate (PO₄) ≤2 ppm
Sulfate (SO₄) ≤2 ppm
Trace Impurities - Aluminum (Al) ≤200.0 ppb
Arsenic and Antimony (as As) ≤3000.0 ppb
Trace Impurities - Barium (Ba) ≤300.0 ppb
Trace Impurities - Boron (B) ≤50 ppb
Trace Impurities - Chromium (Cr) ≤100.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Gold (Au) ≤200.0 ppb
Heavy Metals (as Pb) ≤500.0 ppb
Trace Impurities - Iron (Fe) ≤100.0 ppb
Trace Impurities - Lead (Pb) ≤200.0 ppb
Trace Impurities - Magnesium (Mg) ≤200.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Nickel (Ni) ≤100.0 ppb
Trace Impurities - Tin (Sn) ≤100 ppb
Trace Impurities - Titanium (Ti) ≤100.0 ppb
Trace Impurities - Zinc (Zn) ≤100.0 ppb
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR