Toluene ≥99.5% (by GC), BAKER ANANYZED™ ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

9460-R 9460-03 9460-07 9460-05 9460-33 BAKR9460-01
JT9460-REA 2644.91 USD
JT9460-R JT9460-22 JT9460-3 JT9460-7 JT9460-5 JT9460-33 JT9460-01
Toluene ≥99.5% (by GC), BAKER ANANYZED™ ACS
Toluene

Lot analysis on label.

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₅CH₃) (by GC) ≥99.5 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤0.003 %
Water (by KF, coulometric) ≤0.03 %
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR