N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

Management of Change (MOC) category = N

9344-13
JT9344-13CS 2046.45 USD
JT9344-13
N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®
N,N-Dimethylformamide

For Biotechnology applications.

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: -61
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (HCON(CH₃)₂) (by GC, corrected for water) ≥99.8 %
Appearance Passes Test
Color (APHA) ≤15
Residue after Evaporation ≤5 ppm
Titrable Acid (meq/g) ≤0.0005
Titrable Base (meq/g) ≤0.003
Water (H₂O)(by Karl Fischer titrn) ≤400 ppm
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm ≤0.010
Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 310 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 295 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm ≤0.30
Ultraviolet Absorbance (1.00-cm cell vs. water) - 270 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - Amines (as dimethylamine) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR