TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99 Dried, ultrapure_bioreagent LC, LC-MS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

M.O.C. Category N

4103-01 4103-06 4103-02 4103-F7 4103-04 4103-07 4103-08
JT4103-1EA 392.84 USD
JT4103-1 JT4103-6 JT4103-2 JT4103-F7 JT4103-4 JT4103-7 JT4103-08
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99 Dried, ultrapure_bioreagent LC, LC-MS, J.T.Baker®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)

For liquid chromatography and molecular biology applications.


  • Lot analysis on label

Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Melting Pt: 150…152 °C
Density: 1.05 g/ml (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NH₂C(CH₂OH)₃·HCl) (dried basis) ≥99.0 %
Appearance Passes Test
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Trace Impurities - Calcium (Ca) ≤1 ppm
Trace Impurities - Copper (Cu) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤1 ppm
Trace Impurities - Magnesium (Mg) ≤1.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR