Disodium hydrogen phosphate 98 Dried USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

M.O.C. Category HR

3827-01 3827-07 3827-19 3827-R 3827-09
JT3827-1CS 1138.57 USD
JT3827-1 JT3827-7 JT3827-19 JT3827-R JT3827-09
Disodium hydrogen phosphate 98 Dried USP, J.T.Baker®
di-Sodium hydrogen phosphate

GMP manufactured. Suitable for buffer solutions.


  • Food GMP manufactured product

Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤ 3 ppm
USP - Chloride (Cl) ≤ 0.06 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Matter ≤ 0.2 %
USP - Loss on Drying at 130°C ≤ 5.0 %
USP - Sulfate (SO₄) ≤ 0.2 %
(Suitable for Buffer Solutions)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR