Sodium acetate trihydrate ≥99% (dried basis), crystals USP, Multi-Compendial for biochemistry, for chromatography, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Acetic acid sodium salt trihydrate

Management of Change (MOC) category = HR

3461-07 3461-05 3461-01 3461-08 3461-10 3461-19 3461-06
JT3461-7EA 1596.42 USD
JT3461-7 JT3461-5 JT3461-1 JT3461-8 JT3461-10 JT3461-19 JT3461-06
Sodium acetate trihydrate ≥99% (dried basis), crystals USP, Multi-Compendial for biochemistry, for chromatography, J.T.Baker®
Sodium acetate trihydrate

Optimize biopharmaceutical fermentation and cell culture productivity and improve the efficiency of your pharmaceutical production and biopharmaceutical processes with our proven-effective vitamins and pharmaceutical process minerals.


  • Food GMP manufactured product

Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58
Density: 1.45 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
Meets F.C.C. Requirements
Food GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
Preserve in tight containers, store at room temperature.
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
USP - Calcium and Magnesium Passes Test
USP - Chloride (Cl) ≤ 0.035 %
USP - Endotoxin Concentration (EU/g) ≤ 2.5
USP - Identification Passes Test
USP - Insoluble Matter ≤ 0.05 %
USP - Loss on Drying 38.0 - 41.0 %
USP - pH of 5% Solution at 25°C 7.5 - 9.2
USP - Potassium (K) Passes Test
USP - Sulfate (SO₄) ≤ 0.005 %
EP/BP - Assay (CH₃COONa) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH (1 in 20) 7.5 - 9.0
EP/BP - Reducing Substances Passes Test
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 200 ppm
EP/BP - Calcium and Magnesium ≤ 50 ppm
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying 39.0 - 40.5 %
JP - Assay (CH₃COONa) 99.5 - 101.0 %
JP - Identification Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Chloride (Cl) ≤ 0.011 %
JP - Sulfate (SO₄) ≤ 0.017 %
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Calcium and Magnesium Passes Test
JP - Arsenic (As) ≤ 2 ppm
JP - Substances Reducing Permanganate Passes Test
JP - Loss on Drying 39.0 - 40.5 %
FCC - Alkalinity ≤ 0.05 %
FCC - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Loss on Drying 38.0 - 41.0 %
FCC - Potassium Compounds Passes Test
No Class 1, 2, 3 or other solvent is likely to be present in the final
product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR