Potassium permanganate 99, BAKER ANANYZED™ ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Permanganic acid potassium salt

M.O.C. Category N

3228-07
JT3228-7EA 1337.73 USD
JT3228-7
Potassium permanganate 99, BAKER ANANYZED™ ACS
Potassium permanganate

Crystals. Lot analysis on label.

Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KMnO₄) ≥99.0 %
Insoluble Matter ≤0.2 %
Chloride and Chlorate (as Cl) ≤0.005 %
Sulfate (SO₄) ≤0.02 %
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR