1-Hexanesulfonic acid sodium salt 98,0 HPLC, Ionpair, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate

M.O.C. Category N

2175-05 2175-01
JT2175-5EA 284.51 USD
JT2175-5 JT2175-1
1-Hexanesulfonic acid sodium salt 98,0 HPLC, Ionpair, J.T.Baker®
1-Hexanesulphonic acid sodium salt

Crystals. For ion-pair chromatography of basic compounds.

Formula: C₆H₁₃O₃SNa
MW: 188.22 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as C₆H₁₃SO₃Na) (by acidimetry) ≥98.0 %
UV Absorbance of 0.25 M Solution - 250 nm ≤0.050
UV Absorbance of 0.25 M Solution - 240 nm ≤0.050
UV Absorbance of 0.25 M Solution - 230 nm ≤0.050
UV Absorbance of 0.25 M Solution - 220 nm ≤0.060
UV Absorbance of 0.25 M Solution - 210 nm ≤0.080
UV Absorbance of 0.25 M Solution - 200 nm ≤0.200
Appearance (clear, colorless, mobile liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR